accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 330641-16-2

IUPAC Name: tetrafluoroboranuide; {[(6-chloro-1H-1,2,3-benzotriazol-1-yl)oxy](dimethylamino)methylidene}dimethylazanium
Synonyms: {[(6-chloro-1,2,3-benzotriazol-1-yl)oxy](dimethylamino)methylidene}dimethylazanium tetrafluoroborate
Molecular Weight (g/mol): 355.53
Molecular Formula: C11H15BClF4N5O
InChi Key: GBGVQFJZGHBZMC-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CN(C)C(ON1N=NC2=CC=C(Cl)C=C12)=[N+](C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results