accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 330793-45-8

BOHNHOOH

CAS RN 330793-45-8

IUPAC Name: [4-(phenylcarbamoyl)phenyl]boronic acid
Synonyms: 4-(phenylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 241.05
Molecular Formula: C13H12BNO3
InChi Key: DAUSGSRIYCUJDK-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results