accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOHHO(Z)

CAS RN 331-39-5

IUPAC Name: (2Z)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
Synonyms: caffeicacid ccris 847 caffeic acid pure
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: QAIPRVGONGVQAS-RQOWECAXSA-N
SMILES: OC(=O)\C=C/C1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (5)
  • (13)
Quantity 
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (5)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (3)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (5)
  • (5)
  • (13)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)