accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 33125-05-2

CH3H3CCH3OOHNOOH

CAS RN 33125-05-2

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-2-phenylacetic acid
Synonyms: [(tert-butoxycarbonyl)amino](phenyl)acetic acid
Molecular Weight (g/mol): 251.28
Molecular Formula: C13H17NO4
InChi Key: HOBFSNNENNQQIU-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(C(O)=O)C1=CC=CC=C1
Quantity 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)