accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 331256-09-8

HNSOONOONOO

CAS RN 331256-09-8

IUPAC Name: 2-nitro-N-(2-nitrophenyl)benzene-1-sulfonamide
Synonyms: 2-nitro-N-(2-nitrophenyl)benzenesulfonamide
Molecular Weight (g/mol): 323.28
Molecular Formula: C12H9N3O6S
InChi Key: NGHQCODQNBAKLD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1NS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results