accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOOH3COCH3(E)

CAS RN 33130-03-9

IUPAC Name: (2E)-3-(2,3,4-trimethoxyphenyl)prop-2-enoate
Synonyms: (2E)-3-(2,3,4-trimethoxyphenyl)prop-2-enoate
Molecular Weight (g/mol): 237.23
Molecular Formula: C12H13O5
InChi Key: ZYOPDNLIHHFGEC-FNORWQNLSA-M
SMILES: COC1=CC=C(\C=C\C([O-])=O)C(OC)=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results