accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 331434-68-5

IUPAC Name: N-[(4-fluorophenyl)methyl]-4-nitrobenzamide
Synonyms: N-[(4-fluorophenyl)methyl]-4-nitrobenzamide
Molecular Weight (g/mol): 274.25
Molecular Formula: C14H11FN2O3
InChi Key: JNVXYLPECATQFV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C(=O)NCC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results