accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3316-09-4

OHHONOO

CAS RN 3316-09-4

IUPAC Name: 4-nitrobenzene-1,2-diol
Synonyms: 4-nitrocatechol
Molecular Weight (g/mol): 155.11
Molecular Formula: C6H5NO4
InChi Key: XJNPNXSISMKQEX-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)