accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaNaOPOOOHN

CAS RN 3318-43-2

IUPAC Name: disodium 1H-indol-3-yl phosphate
Synonyms: disodium 1H-indol-3-yl phosphate
Molecular Weight (g/mol): 257.09
Molecular Formula: C8H6NNa2O4P
InChi Key: WCSKWBKPKXJWEG-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]P([O-])(=O)OC1=CNC2=CC=CC=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results