accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 331942-47-3

OOOH

CAS RN 331942-47-3

IUPAC Name: 2-(furan-2-yl)benzoic acid
Synonyms: 2-(furan-2-yl)benzoic acid
Molecular Weight (g/mol): 188.18
Molecular Formula: C11H8O3
InChi Key: QRUHYAWZHFTNEA-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results