accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 332-43-4

FCl

CAS RN 332-43-4

IUPAC Name: 1-(2-chloroethyl)-4-fluorobenzene
Synonyms: 1-(2-chloroethyl)-4-fluorobenzene
Molecular Weight (g/mol): 158.6
Molecular Formula: C8H8ClF
InChi Key: LPUURRVXQCVXCX-UHFFFAOYSA-N
SMILES: FC1=CC=C(CCCl)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)