accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3320-86-3

IUPAC Name: 1-isocyanato-2-nitrobenzene
Synonyms: benzene, 1-isocyanato-2-nitro-
Molecular Weight (g/mol): 164.12
Molecular Formula: C7H4N2O3
InChi Key: JRVZITODZAQRQM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1N=C=O
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)