accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3321-64-0

IUPAC Name: butylphosphonic acid
Synonyms: butylphosphonate
Molecular Weight (g/mol): 138.1
Molecular Formula: C4H11O3P
InChi Key: UOKRBSXOBUKDGE-UHFFFAOYSA-N
SMILES: CCCCP(O)(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results