accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COSOOFFF

CAS RN 333-27-7

IUPAC Name: methyl trifluoromethanesulfonate
Synonyms: triflate ester
Molecular Weight (g/mol): 164.1
Molecular Formula: C2H3F3O3S
InChi Key: OIRDBPQYVWXNSJ-UHFFFAOYSA-N
SMILES: COS(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results