accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FSOOCl

CAS RN 33330-46-0

IUPAC Name: 1-(2-chloroethanesulfonyl)-4-fluorobenzene
Synonyms: 1-(2-chloroethanesulfonyl)-4-fluorobenzene
Molecular Weight (g/mol): 222.66
Molecular Formula: C8H8ClFO2S
InChi Key: VOJIERKKDNYFCX-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)S(=O)(=O)CCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results