accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 333338-18-4

IUPAC Name: disodium hexahydrate 4-nitrophenyl phosphate
Synonyms: disodium hexahydrate p-nitrophenyl phosphate
Molecular Weight (g/mol): 371.14
Molecular Formula: C6H16NNa2O12P
InChi Key: KAKKHKRHCKCAGH-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results