accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NINH2

CAS RN 33348-34-4

IUPAC Name: 4-amino-3-iodobenzonitrile
Synonyms: 4-amino-3-iodobenzonitrile
Molecular Weight (g/mol): 244.04
Molecular Formula: C7H5IN2
InChi Key: UOWVTQFTEAYDLM-UHFFFAOYSA-N
SMILES: NC1=C(I)C=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results