accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 334-48-5

IUPAC Name: decanoic acid
Synonyms: capric acid c10:0 aliphatic carboxylic acid epa pesticide chemical code 128955 sodium salt decanoic acid N-capric acid C10 fatty acid dekansaeure econosan acid sanitizer n-capric acid decanoic acid (natural)
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: GHVNFZFCNZKVNT-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (9)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results