accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOOOOOHFe

CAS RN 334024-15-6

IUPAC Name: iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate hydrate
Synonyms: iron(3+) citrate hydrate
Molecular Weight (g/mol): 262.96
Molecular Formula: C6H7FeO8
InChi Key: AJVRSHNXSHMMCH-UHFFFAOYSA-K
SMILES: O.[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results