accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NONOO

CAS RN 3345-50-4

IUPAC Name: 2,5-dioxo-2,5-dihydro-1H-pyrrole-1-carboxamide
Synonyms: 2,5-dioxopyrrole-1-carboxamide
Molecular Weight (g/mol): 140.1
Molecular Formula: C5H4N2O3
InChi Key: BNPFHEFZJPVCCE-UHFFFAOYSA-N
SMILES: NC(=O)N1C(=O)C=CC1=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results