accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NHNFFF

CAS RN 33468-69-8

IUPAC Name: 5-(trifluoromethyl)-1H-imidazole
Synonyms: 4-(trifluoromethyl)-3H-imidazole
Molecular Weight (g/mol): 136.08
Molecular Formula: C4H3F3N2
InChi Key: DFLGRTIPTPCKPJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=CN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results