accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFFClClClFFF

CAS RN 335-44-4

IUPAC Name: 2,2,3-trichloro-1,1,1,3,4,4,4-heptafluorobutane
Synonyms: 2,2,3-trichloro-1,1,1,3,4,4,4-heptafluorobutane
Molecular Weight (g/mol): 287.38
Molecular Formula: C4Cl3F7
InChi Key: ZPGMWBFCBUKITA-UHFFFAOYNA-N
SMILES: FC(F)(F)C(F)(Cl)C(Cl)(Cl)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results