accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3ClO

CAS RN 3350-78-5

IUPAC Name: 3-methylbut-2-enoyl chloride
Synonyms: 3-methyl-but-2-enoyl chloride
Molecular Weight (g/mol): 118.56
Molecular Formula: C5H7ClO
InChi Key: BDUBTLFQHNYXPC-UHFFFAOYSA-N
SMILES: CC(C)=CC(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results