accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3351-28-8

H3C

CAS RN 3351-28-8

IUPAC Name: 1-methylchrysene
Synonyms: 1-methylchyrsene
Molecular Weight (g/mol): 242.32
Molecular Formula: C19H14
InChi Key: LQKGSLMFGWWLIU-UHFFFAOYSA-N
SMILES: CC1=C2C=CC3=C(C=CC4=CC=CC=C34)C2=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results