accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 33527-91-2

CH3NH3CNNCH3H3CNCH3CH3

CAS RN 33527-91-2

IUPAC Name: (2-{bis[2-(dimethylamino)ethyl]amino}ethyl)dimethylamine
Synonyms: (2-{bis[2-(dimethylamino)ethyl]amino}ethyl)dimethylamine
Molecular Weight (g/mol): 230.4
Molecular Formula: C12H30N4
InChi Key: VMGSQCIDWAUGLQ-UHFFFAOYSA-N
SMILES: CN(C)CCN(CCN(C)C)CCN(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results