accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHNOBrBr

CAS RN 33549-66-5

IUPAC Name: 4,4-dibromo-3-methyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 4,4-dibromo-5-methyl-2H-pyrazol-3-one
Molecular Weight (g/mol): 255.9
Molecular Formula: C4H4Br2N2O
InChi Key: QQQQLBQGDDSBCA-UHFFFAOYSA-N
SMILES: CC1=NNC(=O)C1(Br)Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results