CAS RN 33570-04-6
IUPAC Name:
(1S,4R,7R,8S,9R,11S)-9-tert-butyl-7,9-dihydroxy-3,5,12-trioxatetracyclo[6.6.0.0¹,¹¹.0⁴,⁸]tetradecane-2,6,13-trione
Synonyms:
bilobalide
Molecular Weight (g/mol):
326.3
Molecular Formula:
C15H18O8
InChi Key:
MOLPUWBMSBJXER-YDGSQGCISA-N
SMILES:
CC(C)(C)[C@]1(O)C[C@@H]2OC(=O)C[C@@]22C(=O)O[C@@H]3OC(=O)[C@H](O)[C@]123
1
–
3
of
3
results
1
Bilobalide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Bilobalide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Bilobalide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.