accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

I

CAS RN 33574-02-6

IUPAC Name: (iodomethyl)cyclopropane
Synonyms: (iodomethyl)cyclopropane
Molecular Weight (g/mol): 182
Molecular Formula: C4H7I
InChi Key: JHKJQWFHNIOUKY-UHFFFAOYSA-N
SMILES: ICC1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results