accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 336100-46-0

IUPAC Name: 1H-pyrrole-3-carboxylate
Synonyms: 1H-pyrrole-3-carboxylate
Molecular Weight (g/mol): 110.09
Molecular Formula: C5H4NO2
InChi Key: DOYOPBSXEIZLRE-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CNC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results