CAS RN 33632-27-8
IUPAC Name:
1-(5-nitro-2,3-dihydro-1H-indol-1-yl)ethan-1-one
Synonyms:
1-(5-nitro-2,3-dihydroindol-1-yl)ethanone
Molecular Weight (g/mol):
206.2
Molecular Formula:
C10H10N2O3
InChi Key:
OJKKTNFCWMBYMT-UHFFFAOYSA-N
SMILES:
CC(=O)N1CCC2=CC(=CC=C12)[N+]([O-])=O
1
–
1
of
1
results
1
1-Acetyl-5-nitroindoline, 98%, Thermo Scientific™
CAS: 33632-27-8 Molecular Formula: C10H10N2O3 Molecular Weight (g/mol): 206.20 MDL Number: MFCD00005811 InChI Key: OJKKTNFCWMBYMT-UHFFFAOYSA-N Synonym: 1-acetyl-5-nitroindoline,1-5-nitroindolin-1-yl ethanone,1h-indole, 1-acetyl-2,3-dihydro-5-nitro,indoline, 1-acetyl-5-nitro,ethanone, 1-2,3-dihydro-5-nitro-1h-indol-1-yl,1-5-nitro-2,3-dihydroindol-1-yl ethanone,1-5-nitro-2,3-dihydro-1h-indol-1-yl ethan-1-one,acmc-1ahhj,5-nitro-n-acetylindoline,maybridge1_006268 PubChem CID: 36566 SMILES: CC(=O)N1CCC2=CC(=CC=C12)[N+]([O-])=O