accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 33636-93-0

IUPAC Name: hexakis(triphenylphosphane) hexacopper
Synonyms: stryker's reagent
Molecular Weight (g/mol): 1955.03
Molecular Formula: C108H90Cu6P6
InChi Key: KVBHFBAACGHMKE-UHFFFAOYSA-N
SMILES: [Cu].[Cu].[Cu].[Cu].[Cu].[Cu].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results