accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3OHO(E)

CAS RN 3368-21-6

IUPAC Name: (2E)-3-[4-(propan-2-yl)phenyl]prop-2-enoic acid
Synonyms: (2E)-3-(4-isopropylphenyl)prop-2-enoic acid
Molecular Weight (g/mol): 190.24
Molecular Formula: C12H14O2
InChi Key: SJDOOXOUSJDYFE-VMPITWQZSA-N
SMILES: CC(C)C1=CC=C(\C=C\C(O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results