CAS RN 336879-53-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Magnesium gluconate hydrate
CAS: 336879-53-9 Molecular Formula: C12H24MgO15 Molecular Weight (g/mol): 432.61 MDL Number: MFCD00150971 InChI Key: IAKLPCRFBAZVRW-XRDLMGPZSA-L IUPAC Name: magnesium(2+) 2,3,4,5,6-pentahydroxyhexanoate SMILES: O.[Mg++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O