accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 33763-20-1

CH3NSOO

CAS RN 33763-20-1

IUPAC Name: 4-methyl-2-phenyl-1,3-thiazole-5-carboxylate
Synonyms: 4-methyl-2-phenyl-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 218.25
Molecular Formula: C11H8NO2S
InChi Key: CRSMRBYEBHOYRM-UHFFFAOYSA-M
SMILES: CC1=C(SC(=N1)C1=CC=CC=C1)C([O-])=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)