accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3H2N

CAS RN 3378-72-1

IUPAC Name: benzyl(tert-butyl)azanium
Synonyms: benzyl(tert-butyl)azanium
Molecular Weight (g/mol): 164.27
Molecular Formula: C11H18N
InChi Key: DLSOILHAKCBARI-UHFFFAOYSA-O
SMILES: CC(C)(C)[NH2+]CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results