CAS RN 3378-82-3
IUPAC Name:
1-bromonaphthalene-2-carbaldehyde
Synonyms:
1-bromonaphthalene-2-carbaldehyde
Molecular Weight (g/mol):
235.08
Molecular Formula:
C11H7BrO
InChi Key:
CYGUXEZVBLMVRV-UHFFFAOYSA-N
SMILES:
BrC1=C2C=CC=CC2=CC=C1C=O
1
–
3
of
3
results
1
1-Bromo-2-naphthaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Bromo-2-naphthaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-Bromo-2-naphthaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.