accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 33791-58-1

OOCH2X

CAS RN 33791-58-1

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 205.28
Molecular Formula: C13H17O2
InChi Key:
SMILES: *OC(=O)C=C.C1CC2CC1C1CC=CC21
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results