CAS RN 337958-60-8
IUPAC Name:
5,7-dichloro-1,6-naphthyridine
Synonyms:
5,7-dichloro-1,6-naphthyridine
Molecular Weight (g/mol):
199.03
Molecular Formula:
C8H4Cl2N2
InChi Key:
HNRPBMNTCYRAJD-UHFFFAOYSA-N
SMILES:
ClC1=CC2=NC=CC=C2C(Cl)=N1
1
–
1
of
1
results
1
5,7-Dichloro-1,6-naphthyridine, 97%
CAS: 337958-60-8 Molecular Formula: C8H4Cl2N2 Molecular Weight (g/mol): 199.04 InChI Key: HNRPBMNTCYRAJD-UHFFFAOYSA-N Synonym: pubchem14653,5,7-dichloro-1,6-napthyridine,1,6-naphthyridine,5,7-dichloro,1,6-naphthyridine, 5,7-dichloro,5,7-bis chloranyl-1,6-naphthyridine PubChem CID: 12204233 IUPAC Name: 5,7-dichloro-1,6-naphthyridine SMILES: C1=CC2=C(N=C(C=C2N=C1)Cl)Cl