accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3CCH3OH

CAS RN 337966-85-5

IUPAC Name: 1-cyclopentyl-2,2-dimethylpropan-1-ol
Synonyms: 1-cyclopentyl-2,2-dimethylpropan-1-ol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: BCTWRMZVIPKMLP-UHFFFAOYNA-N
SMILES: CC(C)(C)C(O)C1CCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results