CAS RN 3380-30-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diclosan, TRC
CAS: 3380-30-1 Molecular Formula: C12 H8 Cl2 O2 Molecular Weight (g/mol): 255.1 Synonym: Phenol, 5-chloro-2-(p-chlorophenoxy)- (7CI,8CI),5-Chloro-2-(4-chlorophenoxy)phenol,2-Hydroxy-4,4'-dichlorodiphenyl ether,4,4'-Dichloro-2-hydroxydiphenyl ether,Diclosan,Soneclosan,Tinopal HP 100,Tinosan HP 100,p-Chlorophenyl 2-hydroxy-4-chlorophenyl ether IUPAC Name: 5-chloro-2-(4-chlorophenoxy)phenol SMILES: Oc1cc(Cl)ccc1Oc2ccc(Cl)cc2