accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3381-26-8

H3COHCH3

CAS RN 3381-26-8

IUPAC Name: henicosan-11-ol
Synonyms: henicosan-11-ol
Molecular Weight (g/mol): 312.58
Molecular Formula: C21H44O
InChi Key: BCNCKKYOXRHQGT-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC(O)CCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results