accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaH3CNH3CCH3OPOOOPOOO(R)O(R)(R)OH(S)HONNONH2

CAS RN 33818-15-4

IUPAC Name: sodium 2-(trimethylazaniumyl)ethyl ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Synonyms: sodium cdp-choline(1-)
Molecular Weight (g/mol): 510.31
Molecular Formula: C14H25N4NaO11P2
InChi Key: YWAFNFGRBBBSPD-OCMLZEEQSA-M
SMILES: [Na+].C[N+](C)(C)CCOP([O-])(=O)OP([O-])(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(N)=NC1=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results