accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClH3CNCH3CClCl

CAS RN 33842-02-3

IUPAC Name: dichloro(dimethylamino)methylium chloride
Synonyms: dichloro(dimethylamino)methylium chloride
Molecular Weight (g/mol): 162.44
Molecular Formula: C3H6Cl3N
InChi Key: NRNFKRFWZQQDMD-UHFFFAOYSA-M
SMILES: [Cl-].CN(C)[C+](Cl)Cl
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)