accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 338453-21-7

IUPAC Name: oxane-4-sulfonyl chloride
Synonyms: oxane-4-sulfonyl chloride
Molecular Weight (g/mol): 184.63
Molecular Formula: C5H9ClO3S
InChi Key: QQSBPTQNEOJFBO-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1CCOCC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results