accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3386-42-3

IUPAC Name: 1,4-dichloro-2-isothiocyanatobenzene
Synonyms: 1,4-dichloro-2-isothiocyanatobenzene
Molecular Weight (g/mol): 204.07
Molecular Formula: C7H3Cl2NS
InChi Key: JHTPBGFVWWSHDL-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)C(=C1)N=C=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results