accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3387-36-8

IUPAC Name: disodium [(2R,3S,4S,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Synonyms: disodium uridine phosphate
Molecular Weight (g/mol): 368.15
Molecular Formula: C9H11N2Na2O9P
InChi Key: KURVIXMFFSNONZ-WAJQJQIOSA-L
SMILES: [Na+].[Na+].O[C@H]1[C@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=CC(=O)NC1=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)