accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 339268-21-2

KKKRuNNNNNNK

CAS RN 339268-21-2

IUPAC Name: λ²-ruthenium(2+) tetrapotassium hexakis(iminomethanide) hydrate
Synonyms: λ²-ruthenium(2+) tetrapotassium hexakis(cyanide) hydrate
Molecular Weight (g/mol): 431.59
Molecular Formula: C6H2K4N6ORu
InChi Key: ZJQKAKIKDOERFU-UHFFFAOYSA-N
SMILES: O.[K+].[K+].[K+].[K+].[Ru++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results