accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SNHHO

CAS RN 3394-05-6

IUPAC Name: (3-hydroxyphenyl)thiourea
Synonyms: 3-hydroxyphenylthiourea
Molecular Weight (g/mol): 168.21
Molecular Formula: C7H8N2OS
InChi Key: BHJYKFUCQNISJA-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=CC(O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results