CAS RN 33941-15-0
IUPAC Name:
1,4,7,10,13-pentaoxa-16-azacyclooctadecan-16-ium
Synonyms:
1,4,7,10,13-pentaoxa-16-azacyclooctadecan-16-ium
Molecular Weight (g/mol):
264.34
Molecular Formula:
C12H26NO5
InChi Key:
NBXKUSNBCPPKRA-UHFFFAOYSA-O
SMILES:
C1COCCOCCOCCOCCOCC[NH2+]1
1
–
1
of
1
results
1
1-Aza-18-crown-6, 95%
CAS: 33941-15-0 Molecular Formula: C12H25NO5 Molecular Weight (g/mol): 263.334 MDL Number: MFCD00075466 InChI Key: NBXKUSNBCPPKRA-UHFFFAOYSA-N PubChem CID: 118578 IUPAC Name: 1,4,7,10,13-pentaoxa-16-azacyclooctadecane SMILES: C1COCCOCCOCCOCCOCCN1