CAS RN 339527-86-5
IUPAC Name:
nickel(2+) dipotassium tetrakis(iminomethanide) hydrate
Synonyms:
nickel(2+) dipotassium tetrakis(cyanide) hydrate
Molecular Weight (g/mol):
258.98
Molecular Formula:
C4H2K2N4NiO
InChi Key:
VQZJEIWYJQFLFX-UHFFFAOYSA-N
SMILES:
O.[K+].[K+].[Ni++].[C-]#N.[C-]#N.[C-]#N.[C-]#N
1
–
1
of
1
results
1
Potassium tetracyanonickelate(II) hydrate
CAS: 339527-86-5 Molecular Formula: C4K2N4Ni Molecular Weight (g/mol): 240.96 MDL Number: MFCD00049669 InChI Key: QOOHKAMDWOKKBR-UHFFFAOYSA-N Synonym: potassium tetracyanonickelate ii hydrate,c4n4ni.2k.h2o,dipotassium tetracyanonickelate 2-hydrate IUPAC Name: nickel(2+) dipotassium tetrakis(iminomethanide) hydrate SMILES: [K+].[K+].N#C[Ni--](C#N)(C#N)C#N